Learn More
Siglec-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke:  Novus Biologicals NBP2-27070
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18167613
  • 361.00 € / 0.10 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Siglec-9 Polyclonal specifically detects Siglec-9 in Bovine, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Siglec-9
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9
Rabbit
Protein A purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human, dwarf chimpanzee, common chimpanzee, great ape, 89% in pig and dog, 84% in cow, rhesus monkey, and horse, and 74% homologous in orangutan.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 2-4ug/ml
Q9Y336
SIGLEC9
A portion of amino acids 40-90 of human Siglec9 was used as the immunogen.
0.1 mg
Cardiovascular Biology
27180
Bovine, Canine, Equine, Monkey, Primate