missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 361.00 €
Spezifikation
| Antigen | Siglec-9 |
|---|---|
| Konzentration | 0.5 mg/mL |
| Verdünnung | Western Blot 2-4ug/ml |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18160175
|
Novus Biologicals
NBP2-27070SS |
0.025 mg |
213.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18167613
|
Novus Biologicals
NBP2-27070 |
0.1 mg |
361.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Siglec-9 Polyclonal specifically detects Siglec-9 in Bovine, Canine, Equine, Monkey, Primate samples. It is validated for Western Blot.Spezifikation
| Siglec-9 | |
| Western Blot 2-4ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Canine, Equine, Monkey, Primate | |
| Q9Y336 | |
| 27180 | |
| A portion of amino acids 40-90 of human Siglec9 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CD329, CDw329, FOAP-9, OBBP-LIKE, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9 | |
| SIGLEC9 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, dwarf chimpanzee, common chimpanzee, great ape, 89% in pig and dog, 84% in cow, rhesus monkey, and horse, and 74% homologous in orangutan. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts