missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-F Rat anti-Mouse, BV421, Clone: E50-2440, BD Horizon™
Rat Monoclonal Antibody
Marke: BD Biosciences 565934
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow cytometry.Spezifikation
Siglec-F | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
E50-2440 | |
Brilliant Violet 421 | |
Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5; CD170 | |
Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ, LOU/M IgG2a κ |
For Reseach Use Only.