Learn More
Sirtuin 1/SIRT1 Antibody (9E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals NBP3-26324-50ul
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30039593
  • 343.00 € / 50 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Sirtuin 1/SIRT1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunocytochemistry
9E9
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
A synthesized peptide derived from Human Sirtuin 1/SIRT1 [UniProt Q96EB6]
50 μL
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific
23411
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen