Learn More
SLC30A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA565535
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: RNVEGVEEVHE LHVWQLAGSR IIATAHIKCE DPTSYMEVAK TIKDVFHNHG IHATTIQPEF ASVGSKSSVV PCELACRTQC ALKQCCGTLP QAPSGKDAEK TPAVSISCLE LSNNLEKKPR RTKAENIPAV VIEIKNMPNK QPESS Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.Spezifikation
SLC30A1 | |
Polyclonal | |
Unconjugated | |
Slc30a1 | |
AI839647; C130040I11Rik; Slc30a1; solute carrier family 30 (zinc transporter), member 1; solute carrier family 30 member 1; solute carrier family 30, member 1; zinc transporter 1; ZNT1; ZnT-1; ZnT-1 zinc transporter; ZRC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7779 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6M5 | |
Slc30a1 | |
Recombinant Human SLC30A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only