Learn More
Invitrogen™ SLC6A1 Recombinant Rabbit Monoclonal Antibody (24GB2390)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557260
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SLC6A1 Recombinant Monoclonal Antibody for Western Blot, Flow
GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
Spezifikation
| SLC6A1 | |
| Recombinant Monoclonal | |
| 1.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31648 | |
| SLC6A1 | |
| A synthesized peptide derived from human SLC6A1 / GAT1 (aa 1-50). | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2390 | |
| Unconjugated | |
| SLC6A1 | |
| A730043E01; GABA T1; GABA T-1; GABA transporter 1; GABA transporter protein; GABAT1; GABATHG; GABATR; Gabt; GABT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT1; GAT-1; MAE; SLC6A1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; solute carrier family 6 member 1; XT-1; Xtrp1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232333 | |
| -20°C | |
| Liquid |