Learn More
Ibidi μ-Slide 18 Well - Flat ibiTreat
Artikelnummer. 17276233
Encompass_Preferred
Ansicht ändern
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17276233 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17276233 Lieferant Ibidi Lieferanten-Nr. 81826
Please to purchase this item. Need a web account? Register with us today!
μ-Slide 18 Well - Flat ibiTreat is a chambered coverslip with 18 wells.
  • A chambered coverslip with 18 wells for use in matrix tests and quick immunofluorescence stainings
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Chambered Coverglass
Sterilität Steril