Learn More
Ibidi μ-Slide 2 Well Co-Culture ibiTreat
Artikelnummer. 15573111
Encompass_Preferred
Ansicht ändern
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15573111 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15573111 Lieferant Ibidi Lieferanten-Nr. 81806
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Slide 2 Well Co-Culture ibiTreat is a chambered coverslip with 2 x 9 wells.
  • A chambered coverslip with 2 x 9 wells suitable for co-cultivation assays in combination with microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 2
Sterilität Steril