Learn More
Ibidi μ-Slide 2 Well Uncoated
Beschreibung
- A chambered coverslip with 2 wells for cell culture, immunofluorescence, and high-end microscopy
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Spezifikation
Spezifikation
| Typ | Chambered Coverglass |
| Wells | 2 |
| Sterilität | Steril |