Learn More
Ibidi μ-Slide 8 Well Collagen I
Artikelnummer. 17844834
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17844834 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17844834 Lieferant Ibidi Lieferanten-Nr. 80829
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide 8 Well Collagen I is a chambered coverslip with 8 wells.
  • A chambered coverslip with 8 wells for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • Collagen I coated
  • sterilized
Spezifikation
Typ Microscope Slide
Wellanzahl 8
Menge 15