Learn More
Ibidi μ-Slide 8 Well Glass Bottom, Bulk Pack
Artikelnummer. 17236213
Encompass_Preferred
Change view
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17236213 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17236213 Lieferant Ibidi Lieferanten-Nr. 8082790
Please to purchase this item. Need a web account? Register with us today!
μ-Slide 8 Well Glass Bottom, Bulk Pack is a chambered coverslip with 8 wells.
  • A chambered coverslip with 8 wells for use in TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • individually packed
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8
Sterilität Steril