Learn More
Ibidi μ-Slide 8-Well Chambered Coverslip
Artikelnummer. 17226213
Encompass_Preferred
Ansicht ändern
Click to view available options
Material:
Borosilicate Glass (1.5)
Packungsgröße:
15 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Material unitSize
17226213 Borosilicate Glass (1.5) 15 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17226213 Lieferant Ibidi Lieferanten-Nr. 80827
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Slide 8 Well Glass Bottom is a chambered coverslip with 8 wells.
  • A chambered coverslip with 8 wells for use in TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • Sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8
Material Borosilicate Glass (1.5)
Dicke (metrisch) 170 μm
Sterilität Steril