Learn More
Ibidi μ-Slide 8 Well High Glass Bottom, Bulk Pack
Artikelnummer. 17266203
Encompass_Preferred
Change view
Click to view available options
:
μ-Slide
Artikelnummer.
17266203 μ-Slide
1 options
Artikelnummer. 17266203
Marke: Ibidi 8080790
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Slide 8 Well High Glass Bottom, Bulk Pack is a chambered coverslip with 8 individual wells and high walls.
  • A chambered coverslip with 8 individual wells and high walls
  • suitable for use in TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • individually packed
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8