Learn More
Ibidi μ-Slide 8 Well High ibiTreat
Artikelnummer. 17179985
Encompass_Preferred
Ansicht ändern
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17179985 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17179985 Lieferant Ibidi Lieferanten-Nr. 80806
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Slide 8 Well High ibiTreat is a chambered coverslip with 8 individual wells and high walls.
  • A chambered coverslip with 8 individual wells and high walls for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8