Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD Test Patterns 2
Click to view available options
:
μ-Slide
Beschreibung
- An 8 well chambered coverslip with micropatterned surface to test different patterns for the establishment of cell culture assays
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Spezifikation
Spezifikation
| Typ | Microscope Slide |
| Wellanzahl | 8 |
| Menge | 2 |