Learn More
Ibidi μ-Slide Chemotaxis Collagen IV
Artikelnummer. 17206173
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17206173 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17206173 Lieferant Ibidi Lieferanten-Nr. 80322
 Eingeschränkter Artikel
Verfügbar ab: 23-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide Chemotaxis Collagen IV is a ready-to-use microscopy slide for chemotaxis assays.
  • A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
  • suitable for 2D or 3D cell culture
  • stable chemotactic gradients for long-term experiments
  • bottom material: #1.5 Polymer Coverslip
  • Collagen IV coated
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 10