Learn More
ibidi μ-Slide I 0.2 Luer Uncoated
Artikelnummer. 16281011
Encompass_Preferred
Ansicht ändern
Click to view available options
15 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
16281011 15 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16281011 Lieferant ibidi Lieferanten-Nr. 80161
Verfügbar ab: 11-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
A μ-Slide with one channel
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • Easy to connect with tubing and pump systems via female Luers
  • Channel height: 200 μm, bottom material: #1.5 Polymer Coverslip
  • Hydrophobic
  • Sterilized
Spezifikation
Menge 15 Pk.