Learn More
Ibidi μ-Slide I 0.4 Collagen IV
Product Code. 17226153
Encompass_Preferred
Change view
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17226153 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17226153 Supplier Ibidi Supplier No. 80172
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A μ-Slide has one channel for applying defined shear stress.
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • easy to connect with tubing and pump systems via female Luers
  • channel height: 400 μm
  • bottom material: #1.5 Polymer Coverslip
  • Collagen IV coated
  • sterilized
Specifications
Typ Microscope Slide
Menge 15