Learn More
Ibidi μ-Slide I 0.4 Luer Glass Bottom
Beschreibung
- A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
- easy to connect with tubing and pump systems via female Luers
- channel height: 400 μm
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Spezifikation
Spezifikation
| Typ | Microscope Slide |
| Menge | 15 |