Learn More
Ibidi μ-Slide I Luer Variety Pack Uncoated
Artikelnummer. 17266213
Encompass_Preferred
Ansicht ändern
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17266213 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17266213 Lieferant Ibidi Lieferanten-Nr. 81101
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide I Luer Variety Pack Uncoated is a variety of μ-Slide I Luer channel slides.
  • Variety of μ-Slide I Luer channel slides with 4 times each channel height
  • specially suited for optimizing experimental conditions in flow applications
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 16