Learn More
ibidi μ-Slide Spheroid Perfusion ibiTreat
Artikelnummer. 30130494
Encompass_Preferred
Ansicht ändern
Click to view available options
Anzahl pro Karton:
120/Cs.
90/Cs.
Anzahl pro Packung:
1 Ea.
8/Tray, 15 Trays
Packungsgröße:
120 Stück
90 Stück
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anzahl pro Karton Anzahl pro Packung Packungsgröße
30130494 90/Cs. 1 Ea. 90 Stück
30130571 120/Cs. 8/Tray, 15 Trays 120 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30130494 Lieferant ibidi Lieferanten-Nr. 8035690
 Eingeschränkter Artikel
Verfügbar ab: 07-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
A perfusable channel μ-Slide with 3 x 7 wells for spheroid culture and high-end imaging.
  • Easy to connect with tubing and pump systems via female Luers
  • Bottom material: No. 1.5 Polymer Coverslip
  • Tissue culture treated
  • Sterilized
  • Ideal for the long-term cultivation and perfusion of 3D spheroids or organoids on the Bioinert surface
  • Flexible preparation for perfusion: spheroids can be transferred or created directly in the wells
  • Imaging chamber with excellent optical quality for high-resolution microscopy
Spezifikation
Typ Chamber Slide
Wells 7
Oberflächenbehandlung ibiTreat
Material Polymer
Anzahl pro Karton 90/Cs.
Anzahl pro Packung 1 Ea.
Sterilität Sterile