Learn More
Ibidi μ-Slide VI 0.1 ibiTreat
Artikelnummer. 17296193
Encompass_Preferred
Ansicht ändern
Click to view available options
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17296193 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17296193 Lieferant Ibidi Lieferanten-Nr. 80666
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide VI 0.1 ibiTreat is a 6 channel slide for parallel flow assays and immunofluorescence.
  • A 6 channel slide for parallel flow assays and immunofluorescence that uses a minimal amount of reagents and medium
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 15