Learn More
Ibidi μ-Slide VI 0.1 Uncoated
Beschreibung
- A 6 channel slide for parallel flow assays and immunofluorescence that uses a minimal amount of reagents and medium
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Spezifikation
Spezifikation
| Typ | Microscope Slide |
| Menge | 15 |