Learn More
ibidi μ-Slide VI 0.4 Bioinert
Beschreibung
- Superior to the standard ultra-low attachment (ULA) surfaces - stable, biologically inert surface for long-term experiments without any cell or biomolecule adhesion
- No. 1.5 polymer coverslip
- Surface passivation with Bioinert
- Sterilized
Spezifikation
Spezifikation
| Typ | Chamber Slide |
| Oberflächenbehandlung | Bioinert |
| Material | Polymer |
| Anzahl pro Karton | 90/Cs. |
| Anzahl pro Packung | 1 Ea. |
| Sterilität | Sterile |