Learn More
Ibidi μ-Slide VI 0.4 ibiTreat
Artikelnummer. 16438933
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
15
Packungsgröße:
15 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
16438933 15 15 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16438933 Lieferant Ibidi Lieferanten-Nr. 80606
Verfügbar ab: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide VI 0.4 ibiTreat is a 6 channel slide for parallel flow assays and immunofluorescence.
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 15