Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD Test Patterns 1
μ-Slide VI 0.4 μ-Pattern RGD Test Patterns 1 is a 6 channel slide with micropatterned surface.
Marke: Ibidi 83651
Weitere Details : Netto-Gewicht : 0.57000kg
Beschreibung
- A 6 channel slide with micropatterned surface to test different patterns for the establishment of cell culture assays
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Spezifikation
Microscope Slide | |
2 |