Learn More
Ibidi μ-Slide VI 0.4 Poly-L-Lysine
Artikelnummer. 17226193
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17226193 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17226193 Lieferant Ibidi Lieferanten-Nr. 80604
 Eingeschränkter Artikel
Verfügbar ab: 24-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Slide VI 0.4 Poly-L-Lysine is a 6 channel slide for parallel flow assays and immunofluorescence.
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • Poly-L-Lysine coated
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 15