Learn More
Ibidi μ-Slide VI - Flat ibiTreat
Artikelnummer. 17266193
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
17266193 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17266193 Lieferant Ibidi Lieferanten-Nr. 80626
 Eingeschränkter Artikel
Verfügbar ab: 23-07-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Slide VI - Flat ibiTreat is a 6 channel slide for short term assays.
  • A 6 channel slide for short term assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Microscope Slide
Menge 15