missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SNX3 Recombinant Protein
Marke: Abnova™ H00008724-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH16863 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
38.17 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MAETVADTRRLITKPQNLNDAYGPPSNFLEIDVSNPQTVGVGRGRFTTYEIRVKTNLPIFKLKESTVRRRYSDFEWLRSELERESKPCLRMTSEARSHGRTWCAQNDEKLFCD | |
Grd19/MCOPS8/MGC17570/SDP3/SNX3A | |
SNX3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
8724 | |
SNX3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SNX3 | |
Human | |
Recombinant | |
Solution |