Learn More
SOCS3 Mouse anti-Human, Unconjugated, Clone: 1G2B12, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66797-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The SOCS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOCS3. This antibody recognizes human, mouse antigen. The SOCS3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| SOCS3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 1G2B12 | |
| Unconjugated | |
| 20 μL | |
| 9021 | |
| IgG1 |