missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, 95%, BLD Pharm™

Product Code. 31091899
Encompass_Preferred
Change view
Click to view available options
Menge:
250 mg
1 g
5 g
10 g
25 g
100 g
500 g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31091899 250 mg
31085901 1 g
31087641 5 g
31082418 500 g
31085863 25 g
31090238 10 g
31087250 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31091899 Supplier BLD Pharm Supplier No. BD116536250mg
 Eingeschränkter Artikel

Dieser Artikel erfordert eine Preisaktualisierung, bevor er bestellt werden kann.

Fordern Sie hier einen Preis für diesen Artikel an.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate
CAS 13408-09-8
Summenformel C3H17Na2O11P
MDL-Nummer MFCD00149083
Menge 250 mg
SMILES O=P([O-])([O-])OC(CO)CO.[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[Na+].[Na+]
Molekulargewicht (g/mol) 306.11
Reinheit (%) 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.