missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-pentynoate, MedChemExpress

Artikelnummer. 16486418
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
16486418 10 mg
16442527 50 mg
16426716 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16486418 Lieferant Medchem Express Lieferanten-Nr. HY15286

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types.

Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types.

Spezifikation

Chemischer Name oder Material Sodium 4-pentynoate
Physikalische Form Solid
CAS 101917-30-0
Farbe White
Menge 10 mg
Hinweise zur Reinheitsqualität Research
Summenformel C5H5NaO2
Löslichkeitsinformationen H2O : 100 mg/mL (832.78 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (138.82 mM; Need ultrasonic)
SMILES C#CCCC(O[Na])=O
Molekulargewicht (g/mol) 120.08
Formelmasse 120.08
Reinheit (%) 98.0%
Güte Research
Mehr anzeigen Weniger anzeigen

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.