missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriumchloridlösung, 0.1M, Honeywell Fluka™
Click to view available options
Menge:
500 mL
1 l
Beschreibung
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Spezifikation
Chemischer Name oder Material | Sodium chloride solution |
Menge | 500 mL |
Strukturformel | NaCl |
UN-Nummer | NONH for all modes of transport |
Beilstein | 3534976 |
Formelmasse | 58.44g/mol |
Concentration or Composition (by Analyte or Components) | 0.1M |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'