Learn More
Novus Biologicals™ Sodium Potassium ATPase Beta 1 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Marke:  Novus Biologicals™ NBP2-54665PEP
Artikelnummer. 18260532
  • 259.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ATP1B1. The Sodium Potassium ATPase Beta 1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Spezifikation
Spezifikation
481
adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1
ATP1B1
100 μl
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54100. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
>80% by SDS-PAGE and Coomassie blue staining
Store at -20°C. Avoid freeze-thaw cycles.
35 kDa