Learn More
Splunc2 Monoclonal Antibody (4C7D7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517094
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17094 targets Splunc2 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17094 immunogen is purified recombinant fragment of human Splunc2 (amino acids: 16-250) expressed in E. Coli. MA5-17094 detects Splunc2 which has a predicted molecular weight of approximately 27kDa.
Splunc2 Monoclonal antibody specifically detects Splunc2 in Human samples. It is validated for ELISA, Flow Cytometry, Western Blot.Spezifikation
Splunc2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q96DR5 | |
BPIFA2 | |
Purified recombinant fragment of human Splunc2 (amino acids: 16-250) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
4C7D7 | |
Unconjugated | |
BPIFA2 | |
bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; BPIFA2; Bpifa2e; C20orf70; mSplunc2; parotid secretory protein; PSP; short palate; short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025 | |
Mouse | |
Protein G | |
RUO | |
140683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |