Learn More
Invitrogen™ STK22B (TSSK2) Recombinant Human Protein
STK22B (TSSK2) is a serine/threonine kinase.
Marke: Invitrogen™ PR5947A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
STK22B (TSSK2) is essential for the function and production of sperm (spermiogenesis). Deletions in the STK22B coding region correlate with numerous cardiovascular and glandular malformations.
Spezifikation
NP_443732 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
STK22B (TSSK2) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
45.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
G11 Kinase Family | |
Recombinant | |
Baculovirus |