missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550890
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total SULF1 protein.
SULF1 Polyclonal antibody specifically detects SULF1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
SULF1 | |
Polyclonal | |
Unconjugated | |
SULF1 | |
AI467640; AW121680; extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; Kiaa1077; mKIAA1077; mSulf-1; RSulfFP1; Sulf1; SULF-1; sulfatase 1; sulfatase FP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23213 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IWU6 | |
SULF1 | |
Synthetic peptide corresponding to a region derived from internal residues of human sulfatase 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |