missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO2/SUMO3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA141153
Informations supplémentaires : Netto-Gewicht : 0.01000kg
Description
Suggested positive control: HL-60 or NIH-3T3, HL-60 whole cell lysate.
This gene encodes a protein that is a member of the SUMO (small ubiquitin-like modifier) protein family. It functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. It is not active until the last two amino acids of the carboxy-terminus have been cleaved off. Numerous pseudogenes have been reported for this gene. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Spécification
SUMO2/SUMO3 | |
Polyclonal | |
Unconjugated | |
Sumo3 | |
2810014B19Rik; D10Ertd345e; GMP1; HSMT3; I79_000932; MIF2 suppressor; Sentrin; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 3 L homeolog; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 3; Smt3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3B; SMT3C; SMT3H1; SMT3H2; SMT3H3; SUMO-1; SUMO2; SUMO-2; Sumo3; SUMO-3; sumo3.L; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; UBL1 | |
Rabbit | |
Protein G | |
RUO | |
170930, 20610, 6612, 6613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
P55854, P61956, P61957, Q9Z172 | |
SUMO2, Sumo3 | |
The amino acid sequences used as immunogen for this antibody are 4-16 (EKPKEGVKTENDH) and 61-76 (RFDGQPINETDTPAQL) of human SUMO2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |