Learn More
MedChemExpress SYBR Green qPCR Master Mix (No ROX)
Spezifikation
Spezifikation
| Inhalt und Lagerung | Stored at -20°C protecting from light, and is stable for up to 18 months. Avoid repetitive freeze-thaw cycles while using. For immediate use, components may be stored at 2°C to 8°C. |
| Produkttyp | Kits |
| Menge | 2000 Reactions |
| Versandbedingung | Room temperature |