missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1 Monoclonal Antibody (SP6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517612
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17612 targets Syntaxin-1 in ELISA, FACS, and WB applications and shows reactivity with Human, Mouse, and Rat samples. The MA5-17612 immunogen is human Syntaxin.
Syntaxin 1 Monoclonal antibody specifically detects Syntaxin 1 in Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Syntaxin 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O35526, P32851, Q16623 | |
Stx1a | |
Human Syntaxin | |
200 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
SP6 | |
Unconjugated | |
Stx1a | |
HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2 | |
Mouse | |
Protein G | |
RUO | |
116470, 20907, 6804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |