missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534227
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
Syntaxin 1 | |
Polyclonal | |
Unconjugated | |
Stx1a | |
HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100512594, 100759989, 100855965, 100857565, 20907, 6804, 788566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O35526, P32850, Q16623 | |
Stx1a | |
Synthetic 16 amino acid peptide from internal region of human STX1A / Syntaxin 1A. | |
50 μg | |
Primary | |
Bovine, Canine, Chicken, Hamster, Human, Mouse, Non-human Primate, Porcine | |
Antibody | |
IgG |