Learn More
SYT16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552072
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TRERMMGEKL FYLSHLHPEG EMKVTLVLEP RSNISSGGSP LSPSAVSHSD STSSTQSLSH GGAPELLVGL SYNATTGRLS VEMIKGSHFR NLAVNRAPDT YGKLFLLNSV GQEMSRCKTS IRRGQPNPVY KETFVFQVAL FQLSDVTLM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
SYT16 Polyclonal antibody specifically detects SYT16 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
SYT16 | |
Polyclonal | |
Unconjugated | |
SYT16 | |
chr14 synaptotagmin; CHR14SYT; CTD-2277K2.1; STREP14; Synaptotagmin 14-like protein; synaptotagmin 16; synaptotagmin XIV-related protein; synaptotagmin XVI; synaptotagmin-16; SYT14L; syt14r; SYT16; yt14r | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q17RD7 | |
SYT16 | |
Recombinant protein corresponding to Human SYT16 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only