missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T7 Tag Polyclonal Antibody, HRP, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA131449
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-31449 detects T7 tag from tagged samples.
The T7 Tag is an 11 amino acid peptide derived from the bacteriophage T7 major capsid protein, gene 10. T7 cleavage site: MASMTGGQQMG. Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available.Spezifikation
T7 Tag | |
Polyclonal | |
HRP | |
MASMTGGQQMG; T7 cleavage | |
synthetic peptide, MASMTGGQQMG (T7), conjugated to KLH. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% Pro-Clean 400; pH 7.2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Tag | |
Antibody | |
IgG |