Learn More
TACC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552207
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MSLQVLNDKN VSNEKNTENC DFLFSPPEVT GRSSVLRVSQ KENVPPKNLA KAMKVTFQTP LRDPQTHRIL SPSMASKLEA PFTQDDTLGL ENSHPVWTQK EN Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 75%.
TACC3 Polyclonal antibody specifically detects TACC3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TACC3 | |
Polyclonal | |
Unconjugated | |
TACC3 | |
Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6A5 | |
TACC3 | |
Recombinant protein corresponding to Human TACC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only