Learn More
R&D Systems™ 10X TdT Stop Buffer
Artikelnummer. 30000723
Click to view available options
Menge:
100 ml
Packungsgröße:
100 Milliliter
Artikelnummer. 30000723
Marke: R&D Systems™ 48103003
om dit product te kopen Registreer vandaag om een webaccount aan te maken
R&D Systems™ TACS 2 TdT Stop Buffer is provided with the TACS TdT kits to stop the activity of the TdT enzyme in TUNEL labeling assays.
  • Uncontrolled product according to WHMIS classification criteria
  • All of the ingredients in this product are listed on the United States TSCA (Toxic Substances Control Act) inventory, the Domestic Substances List (DSL) and/or the Non-Domestic Substances List (NDSL)
TRUSTED_SUSTAINABILITY
Spezifikation
Menge 100 ml
Inhalt und Lagerung Store the unopened product at 2°C to 8°C. Do not use past expiration date.