Learn More
TAF15 Monoclonal Antibody (8TA-2B10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3078
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
MA3-078 detects TATA-Binding Protein (TBP) Associated Factor 15 from human, mouse, rat, canine, and non-human primate samples. The MA3-078 immunogen is the RNA- binding region (RNP) of human TAFII68 as a GST-fusion protein (from aa 175 to 414).
TAF15 Monoclonal antibody specifically detects TAF15 in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot.Spezifikation
TAF15 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
Q92804 | |
TAF15 | |
RNA binding region (RNP) of human TAFII68 as a GST-fusion protein (from amino acid 175 to 414) | |
RUO | |
287571, 491135, 70439, 8148 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Immunoprecipitation, Western Blot | |
8TA-2B10 | |
Unconjugated | |
TAF15 | |
2610111C21Rik; 68 kDa TATA-binding protein-associated factor; 68kDa; hTAFII68; LOC100713096; Npl3; RBP56; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; Taf15; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa; TAF2N; TAFII68; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA-binding protein 56); TATA box binding protein associated factor 15; TATA box-binding protein-associated factor 2N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2N; TATA-box binding protein associated factor 15; TBP-associated factor 15 | |
Mouse | |
50 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |
Research Use Only