missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF1C Recombinant Rabbit Monoclonal Antibody (3O8T1)

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen™ MA557997

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 30283570

  • 185.00 € / 20 Mikroliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Immunogen sequence: PWDPGLTARD LLFRGGCRYR KRPRVVLDVT EQISRFLLDH GDVAFAPLGK LMLENFKLEG AGSRTKKKTV VSVKKLLQDL GGHQPWGCPW AYLSNRQRRF SILGGPILGT SVASHLAELL HEELVLRWEQ LLLDEACTGG ALAWVPG.

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

TAF1C
Recombinant Monoclonal
1.04 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Q15572, Q6P773
TAF1C
Recombinant fusion protein containing a sequence corresponding to amino acids 75-221 of human TAF1C (NP_005670.4).
20 μL
Primary
Human, Rat
Antibody
IgG
ELISA, Western Blot
3O8T1
Unconjugated
TAF1C
MGC:39976; mTAFI95; RNA polymerase I-specific TBP-associated factor 110 kDa; RNA polymerase I-specific TBP-associated factor 95 kDa; SL1; SL1, 110kD subunit; TAF1C; TAFI110; TAFI95; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; transcription factor TAFI95; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA box-binding protein-associated factor, RNA polymerase I, subunit C; TATA-box binding protein associated factor, RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; TBP-associated factor RNA polymerase I 95 kDa; transcription factor SL1; transcription factor TAFI95; transcription initiation factor SL1/TIF-IB subunit C
Rabbit
Protein A
RUO
361420, 9013
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Aktionen

Aktionen

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.