Learn More
TAIP12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553822
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FNPIRVRTHY LHTIMKLELE SKRQVSRPAA PDEEPSPTAS CSLTGAQGSE TQDFQEFIAE NETAVMHLQS AEELERLKAE EDSSGSSAS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
The protein encoded by this gene belongs to the CSRNP family of nuclear proteins that share conserved regions, including cysteine- and serine- rich regions, a basic domain, a transcriptional activation domain, and bind the sequence 'AGAGTG', thus have the hallmark of transcription factors. Studies in mice suggest that these genes may have redundant functions. Alternatively spliced transcript variants have been found for this gene.Spezifikation
TAIP12 | |
Polyclonal | |
Unconjugated | |
Csrnp2 | |
BC035295; C12orf2; C12orf22; Csnrp2; CSRN2; Csrnp2; CSRNP-2; cysteine and serine rich nuclear protein 2; cysteine/serine-rich nuclear protein 2; cysteine-serine-rich nuclear protein 2; Fam130a1; family with sequence similarity 130, member A1; FLJ25576; PPP1R72; Protein FAM130A1; protein phosphatase 1, regulatory subunit 72; RGD1308916; TAIP12; TAIP-12; TGF-beta induced apoptosis protein 12; TGF-beta-induced apoptosis protein 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H175 | |
Csrnp2 | |
Recombinant protein corresponding to Human TAIP12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only