Learn More
Invitrogen™ Talacotuzumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558479
 View more versions of this product
Product Code. 30282699
  • 500.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
This item is not returnable. View return policy
Description
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Talacotuzumab, also known as JNJ-56022473 (and formerly CSL-362) is a humanized IgG1 monoclonal antibody designed to bind to and neutralize CD123 (Interleukin-3 receptor alpha chain or IL3RA) with potential antineoplastic activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Talacotuzumab Humanized
Recombinant Monoclonal
Unconjugated
CSL362; CSL-362-AML; JNJ-56022473
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized