missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress TB500

Product Code. 16489378
Encompass_Preferred
Change view
Click to view available options
Menge:
10 mg
Unit Size:
10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16489378 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489378 Supplier MedChemExpress Supplier No. HYP0170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TB500 is a synthetic version of an active region of thymosin β4. TB500 is claimed to promote endothelial cell differentiation, angiogenesis in dermal tissues, keratinocyte migration, collagen deposition and decrease inflammation.

Specifications

Reinheit 98.0%
CAS 885340-08-9
Inhalt und Lagerung Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Zur Verwendung mit (Anwendung) Metabolism-protein/nucleotide metabolism
Molekulargewicht 889.01 g/mol
Produkttyp Peptides
Menge 10 mg
Versandbedingung Room temperature
Löslichkeit H2O : 250 mg/mL (281.21 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.